missing translation for 'onlineSavingsMsg'
Learn More

D-myo-Inositol, 99.2%, Cell culture reagent, For HPLC analysis, MP Biomedicals™

Product Code. 11481881 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
50 g
100 g
1 kg
Unit Size:
100g
1kg
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11481881 1 kg 1kg
11451881 50 g 50g
11461881 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11481881 Supplier MP Biomedicals Supplier No. 0219468891
 Restricted Item
Estimated Shipment: 18-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 87-89-8
Molecular Formula C6H12O6
Molecular Weight (g/mol) 180.156
MDL Number MFCD00077932
InChI Key CDAISMWEOUEBRE-UHFFFAOYSA-N
Synonym scyllo-inositol, myo-inositol, inositol, muco-inositol, epi-inositol, i-inositol, meso-inositol, allo-inositol, 1d-chiro-inositol, myoinositol
PubChem CID 892
ChEBI CHEBI:24848
IUPAC Name cyclohexane-1,2,3,4,5,6-hexol
SMILES C1(C(C(C(C(C1O)O)O)O)O)O

Specifications

Color White
Density 6.2g/mL
Quantity 1 kg
Formula Weight 180.2
Grade Cell Culture Reagent
Physical Form Powder
Chemical Name or Material D-myo-inositol, Molecular biology and Cell biology
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.