missing translation for 'onlineSavingsMsg'
Learn More

D-myo-Inositol, MP Biomedicals™

Product Code. 11398889 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
1 kg
Unit Size:
1kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11398889 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11398889 Supplier MP Biomedicals Supplier No. 0210205291
  • ONLINE EXCLUSIVE PRICE
    649.00 €
    615.30 € / 1kg
    Save 33.70 €
    5% Off
 Restricted Item
Estimated Shipment: 05-03-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Plays an important role as a structural element for secondary messengers in eukaryotic cells. D-myo-Inositol, MP Biomedicals is a growth factor for animals and microorganisms. It serves as an important component of phosphatidylinositol (PI) and the phosph

Myo-Inositol is the most abundant form of polyols that serves as a structural element of secondary messengers in eukaryotic cells.

  • It is a component of membrane phospholipids, glycosyl­phosphatidyl­inositol anchors that bind glycoproteins to cell membranes, and inositol phosphate second messengers
  • Inositol or cyclohexane-1,2,3,4,5,6-hexol is a chemical compound, a sixfold alcohol (polyol) of cyclohexane
  • It exists in nine possible stereoisomers, of which the most prominent form, widely occurring in nature, is cis-1,2,3,5-trans-4,6-cyclohexanehexol, or myo-inositol (former name meso-inositol)
  • Inositol is a carbohydrate, though not a classical sugar. It has a taste which has been assayed at half the sweetness of table sugar (sucrose)

Specifications

CAS 87-89-8
Melting Point 220°C to 230°C
pH Neutral to Litmus (10% aq. soln.)
Molecular Formula C6H12O6
Quantity 1 kg
Synonym scyllo-inositol, myo-inositol, inositol, muco-inositol, epi-inositol, i-inositol, meso-inositol, allo-inositol, 1d-chiro-inositol, myoinositol
Solubility Information Readily soluble in water (50mg/mL); slightly soluble in alcohol; insoluble in ether and other common organic solvents.
InChI Key CDAISMWEOUEBRE-UHFFFAOYSA-N
SMILES C1(C(C(C(C(C1O)O)O)O)O)O
IUPAC Name cyclohexane-1,2,3,4,5,6-hexol
Molecular Weight (g/mol) 180.156
PubChem CID 892
ChEBI CHEBI:24848
Formula Weight 180.2
Physical Form Powder
Show More Show Less
compliance-icons
Product Identifier
  • D-myo-Inositol
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P321-Specific treatment (see on this label).
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: D-myo-Inositol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.