missing translation for 'onlineSavingsMsg'
Learn More

D-ornithine, >99%, MP Biomedicals™

Product Code. 11428140 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
Unit Size:
10g
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11428140 5 g 5g
11418140 1 g 1g
11438140 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11428140 Supplier MP Biomedicals Supplier No. 0210251405
 Restricted Item
Estimated Shipment: 02-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 3184-13-2
Molecular Formula C5H13ClN2O2
Molecular Weight (g/mol) 168.621
InChI Key GGTYBZJRPHEQDG-WCCKRBBISA-N
Synonym l-ornithine hydrochloride, l-ornithine monohydrochloride, h-orn-oh.hcl, l-ornithine hcl, l +-ornithine hydrochloride, l-ornithine, monohydrochloride, s-2,5-diaminopentanoic acid hydrochloride, ornithine hydrochloride van, unii-hbk84k66xh, ornithine l monochlorohydrate
PubChem CID 76654
IUPAC Name (2S)-2,5-diaminopentanoic acid;hydrochloride
SMILES C(CC(C(=O)O)N)CN.Cl

Specifications

Assay Percent Range ∽ 99%
Quantity 5 g
Formula Weight 168.6
Percent Purity >99%
Physical Form Powder
Chemical Name or Material D-Ornithine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.