missing translation for 'onlineSavingsMsg'
Learn More

D-Panthenol, MedChemExpress

Product Code. 16432339
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16432339 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16432339 Supplier Medchem Express Supplier No. HYB1391
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Aggiungi al carrello

missing translation for 'validMainframeMsg'

Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

MedChemExpress D-Panthenol is the biologically-active alcohol of pantothenic acid, which leads to an elevation in the amount of coenzyme A in the cell.

D-Panthenol is the biologically-active alcohol of pantothenic acid, which leads to an elevation in the amount of coenzyme A in the cell.

Specifications

Chemical Name or Material D-Panthenol
CAS 81-13-0
Color Colorless
Purity Grade Notes Research
Molecular Formula C9H19NO4
Quantity 1 g
Synonym Dexpanthenol
Solubility Information Ethanol : 110 mg/mL (535.93 mM; Need ultrasonic) ∣DMSO : 33 mg/mL (160.78 mM; Need ultrasonic and warming)
SMILES O=C(NCCCO)[C@H](O)C(C)(C)CO
Molecular Weight (g/mol) 205.25
Formula Weight 205.25
Percent Purity 98.0%
Grade Research
Physical Form Viscous Liquid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.