missing translation for 'onlineSavingsMsg'
Learn More

D-(-)-Pantolactone, TRC

Product Code. 30420968
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30420968 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30420968 Supplier Toronto Research Chemicals Supplier No. TRCP1825005G

Please to purchase this item. Need a web account? Register with us today!

Este artículo no se puede devolver. Vea la política de devoluciones

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Especificaciones

Chemical Name or Material Pantolactone
CAS 599-04-2
Purity Grade Notes GC
Molecular Formula C6 H10 O3
Quantity 5 g
Synonym D-(-)-Pantolactone2(3H)-Furanone','dihydro-3-hydroxy-4,4-dimethyl-','(3R)-, 2(3H)-Furanone','dihydro-3-hydroxy-4,4-dimethyl-','(R)-, 2(3H)-Furanone','dihydro-3-hydroxy-4,4-dimethyl-','D-(-)- (8CI), (3R)-Dihydro-3-hydroxy-4,4-dimethyl-2(3H)-furanone, (-)-(R)-Pantolactone, (-)-2-Hydroxy-3,3-dimethyl-γ-butyrolactone, (-)-Pantolactone, (-)-Pantoyl lactone, (3R)-Tetrahydro-3-hydroxy-4,4-dimethylfuran-2-one, (R)-(-)-Pantolactone, (R)-3-Hydroxy-4,4-dimethyldihydrofuran-2(3H)-one, (R)-Pantolactone, (R)-α-Hydroxy-β,β-dimethyl-γ-butyrolactone, D-(-)-Pantolactone, D-(-)-Pantoyl lactone, D-(-)-α-Hydroxy-β,β-dimethyl-γ-butyrolactone, D-Pantolactone, D-Pantoyl lactone, Pantolactone, Pantothenic lactone, Pantoyl lactone, l-Pantoyl lactone
InChI Formula InChI=1S/C6H10O3/c1-6(2)3-9-5(8)4(6)7/h4,7H,3H2,1-2H3/t4-/m0/s1
SMILES CC1(C)COC(=O)[C@@H]1O
IUPAC Name (3R)-3-hydroxy-4,4-dimethyloxolan-2-one
Molecular Weight (g/mol) 130.14
Formula Weight 130.063
Percent Purity >95
Mostrar más Mostrar menos
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.