missing translation for 'onlineSavingsMsg'
Learn More

D-Penicillamine Disulfide, TRC

Product Code. 30428325
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30428325 25 g 25g
30409809 1 g -
30422995 5 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30428325 Supplier Toronto Research Chemicals Supplier No. TRCP22299525G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 20902-45-8
Molecular Formula C10 H20 N2 O4 S2
Molecular Weight (g/mol) 296.41
Synonym D-Valine, 3,3'-dithiobis-, Valine, 3,3'-dithiodi-, D- (8CI), D-Penicillamine disulfide, NSC 87505, Oxidized D-penicillamine, Oxidized penicillamine, Penicillamine disulfide
IUPAC Name (2S)-2-amino-3-[[(1S)-1-amino-1-carboxy-2-methylpropan-2-yl]disulfanyl]-3-methylbutanoic acid
SMILES CC(C)([C@H](C(=O)O)N)SSC(C)(C)[C@H](C(=O)O)N

Specifications

Quantity 25 g
InChI Formula InChI=1S/C10H20N2O4S2/c1-9(2,5(11)7(13)14)17-18-10(3,4)6(12)8(15)16/h5-6H,11-12H2,1-4H3,(H,13,14)(H,15,16)/t5-,6-/m0/s1
Formula Weight 296.0864
Chemical Name or Material Penicillamine disulfide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.