missing translation for 'onlineSavingsMsg'
Learn More
Learn More
D-Phenylalaninol, TRC
Click to view available options
Quantity:
1 g
Unit Size:
1g
Specifications
Specifications
| Chemical Name or Material | D-Phenylalaninol |
| CAS | 5267-64-1 |
| Molecular Formula | C9H13NO |
| Quantity | 1 g |
| Synonym | (betaR)-beta-Amino-benzenepropanol, D-2-Amino-3-phenyl-1-propanol, ((1R)-1-Hydroxymethyl-2-phenylethyl)amine, (2R)-2-Amino-3-phenyl-1-propanol, (R)-(+)-2-Amino-3-phenyl-1-glycinol, (R)-2-Amino-1-hydroxy-3-phenylpropane, (R)-2-Benzylethanolamine |
| InChI Formula | InChI=1S/C9H13NO/c10-9(7-11)6-8-4-2-1-3-5-8/h1-5,9,11H,6-7,10H2/t9-/m1/s1 |
| SMILES | N[C@@H](CO)Cc1ccccc1 |
| IUPAC Name | (2R)-2-amino-3-phenylpropan-1-ol |
| Molecular Weight (g/mol) | 151.21 |
| Formula Weight | 151.0997 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction