missing translation for 'onlineSavingsMsg'
Learn More

D-Pinitol, TRC

Product Code. 30355955
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30355955 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30355955 Supplier Toronto Research Chemicals Supplier No. TRCD21294010G
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifica

Chemical Name or Material D-Pinitol
CAS 10284-63-6
Molecular Formula C7 H14 O6
Quantity 10 g
Synonym 3-O-Methyl-D-chiro-inositol, Matezit, Pinit, Sennit, Sennitol
InChI Formula InChI=1S/C7H14O6/c1-13-7-5(11)3(9)2(8)4(10)6(7)12/h2-12H,1H3/t2-,3-,4-,5-,6+,7+/m0/s1
SMILES CO[C@H]1[C@H](O)[C@@H](O)[C@@H](O)[C@H](O)[C@@H]1O
IUPAC Name (1R,2S,4S,5S)-6-methoxycyclohexane-1,2,3,4,5-pentol
Molecular Weight (g/mol) 194.18
Formula Weight 194.079
Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.