missing translation for 'onlineSavingsMsg'
Learn More

D-Proline ethyl ester hydrochloride, 98%

Product Code. 15492319
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15492319 5 g 5g
15402329 25 g 25g
15412329 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15492319 Supplier Thermo Scientific Alfa Aesar Supplier No. H64910.06
 Restricted Item
Estimated Shipment: 02-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 131477-20-8 | C7H14ClNO2 | 179.644 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 131477-20-8
Molecular Formula C7H14ClNO2
Molecular Weight (g/mol) 179.644
MDL Number MFCD00191003
InChI Key DUJGQVVONTYHLT-FYZOBXCZSA-N
Synonym r-ethyl pyrrolidine-2-carboxylate hydrochloride, d-proline ethyl ester hydrochloride, h-d-pro-oet hcl, h-d-pro-oet hydrochloride, d-proline, ethyl ester, hydrochloride, ethyl 2r-pyrrolidine-2-carboxylate hydrochloride, h-d-pro-oet.hcl, h-d-pro-oet . hci, h-d-pro-oet?cl, d-proline ethyl ester hcl
PubChem CID 56777374
IUPAC Name ethyl (2R)-pyrrolidine-2-carboxylate;hydrochloride
SMILES CCOC(=O)C1CCCN1.Cl

Specifications

Color Yellow
Beilstein 7170607
Quantity 5 g
Formula Weight 179.64
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material D-Proline ethyl ester hydrochloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.