missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals D-Propargylglycine, 97%

Product Code. 15412639
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15412639 5 g 5g
15402639 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15412639 Supplier Thermo Scientific Chemicals Supplier No. H65798.06
 Restricted Item
Estimated Shipment: 30-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 23235-03-2 | C5H7NO2 | 113.116 g/mol

A useful biochemical for proteomics research. Also been used for affinity labeling of hog kidney D-amino acid oxidase.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
A useful biochemical for proteomics research. Also been used for affinity labeling of hog kidney D-amino acid oxidase.

Solubility
Slightly soluble in water.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 23235-03-2
Molecular Formula C5H7NO2
Molecular Weight (g/mol) 113.116
MDL Number MFCD00237407
InChI Key DGYHPLMPMRKMPD-SCSAIBSYSA-N
Synonym d-propargylglycine, r-2-aminopent-4-ynoic acid, r-alpha-propargylglycine, unii-2252qef2pl, 4-pentynoic acid, 2-amino-, 2r, 2r-2-aminopent-4-ynoic acid, r-2-propargylglycine, r-2-amino-4-pentynoic acid, d-2-propynylglycine, propargylglycine, d
PubChem CID 6992713
IUPAC Name (2R)-2-aminopent-4-ynoic acid
SMILES C#CCC(C(=O)O)N

Specifications

Melting Point 205°C (decomposition)
Color White
Solubility Information Slightly soluble in water.
Quantity 5 g
Odor Odorless
Formula Weight 113.12
Percent Purity 97%
Physical Form Powder
Chemical Name or Material D-Propargylglycine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.