missing translation for 'onlineSavingsMsg'
Learn More

D-(-)-Quinic acid, 98%

Product Code. 11398216
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11398216 5 g 5g
11308226 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11398216 Supplier Thermo Scientific Alfa Aesar Supplier No. L15238.06
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 77-95-2 | C7H12O6 | 192.17 g/mol

D(-)-Quinic Acid is used in the synthesis of anti-influenza/anti-swine flu medication.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
D(-)-Quinic Acid is used in the synthesis of anti-influenza/anti-swine flu medication.

Solubility
Soluble in water (400g/L), methanol and dimethyl sulfoxide.

Notes
Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 77-95-2
Molecular Formula C7H12O6
Molecular Weight (g/mol) 192.17
MDL Number MFCD00003864
InChI Key AAWZDTNXLSGCEK-UHFFFAOYNA-N
Synonym quinic acid, d---quinic acid, --quinic acid, chinic acid, kinic acid, chinasaure, quinate, l-quinic acid, d-quinic acid, unii-058c04bgyi
PubChem CID 6508
IUPAC Name (3R,5R)-1,3,4,5-tetrahydroxycyclohexane-1-carboxylic acid
SMILES OC1CC(O)(CC(O)C1O)C(O)=O

Specifications

Melting Point 162°C to 167°C
Odor Odorless
Quantity 5 g
Beilstein 2212412
Merck Index 14,8059
Solubility Information Soluble in water (400g/L),methanol and dimethyl sulfoxide.
Optical Rotation −44° (c=20 in Water)
Formula Weight 192.17
Percent Purity 98%
Chemical Name or Material D-(-)-Quinic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.