missing translation for 'onlineSavingsMsg'
Learn More

D(-)-Quinic acid, 98+%

Product Code. 10146130 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10146130 25 g Glass bottle 25g
10659014 100 g Glass bottle 100g
11995071 500 g Glass bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10146130 Supplier Thermo Scientific Acros Supplier No. 160500250
 Restricted Item
In Stock
Add to basket
Edge

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 77-95-2 | C7H12O6 | 192.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 77-95-2
Molecular Formula C7H12O6
Molecular Weight (g/mol) 192.17
MDL Number MFCD00003864
InChI Key AAWZDTNXLSGCEK-UHFFFAOYNA-N
Synonym quinic acid, d---quinic acid, --quinic acid, chinic acid, kinic acid, chinasaure, quinate, l-quinic acid, d-quinic acid, unii-058c04bgyi
PubChem CID 6508
IUPAC Name (3R,5R)-1,3,4,5-tetrahydroxycyclohexane-1-carboxylic acid
SMILES OC1CC(O)(CC(O)C1O)C(O)=O

Specifications

Melting Point 166.0°C to 168.0°C
Color White to Yellow
Ignition Residue 0.1% max.
Loss on Drying 2% max.
Infrared Spectrum Authentic
Assay Percent Range 98+%
Packaging Glass bottle
Linear Formula (HO)4C6H7CO2H
Quantity 25 g
Merck Index 15,8175
Specific Rotation Condition - 44.00 (20.00°C c=11.2,H2O)
Specific Rotation − 44.00
Solubility Information Solubility in water: 400g/L (20°C). Other solubilities: soluble in glacial acetic acid and alcohol
Formula Weight 192.17
Percent Purity 98+%
Physical Form Crystalline Powder
Chemical Name or Material D(-)-Quinic acid
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.