missing translation for 'onlineSavingsMsg'
Learn More

D(-)-Quinic Acid, TRC

Product Code. 30411088
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30411088 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30411088 Supplier Toronto Research Chemicals Supplier No. TRCQ67063010G
 Restricted Item
Estimated Shipment: 21-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Quinic Acid
CAS 77-95-2
Molecular Formula C7 H12 O6
Quantity 10 g
Synonym Cyclohexanecarboxylic acid','1,3,4,5-tetrahydroxy-','(1α,3R,4α,5R)-, Cyclohexanecarboxylic acid','1,3,4,5-tetrahydroxy-','(-)- (8CI), Cyclohexanecarboxylic acid','1,3,4,5-tetrahydroxy-','[1R-(1α,3α,4α,5β)]-, (1α,3R,4α,5R)-1,3,4,5-Tetrahydroxycyclohexanecarboxylic acid, (-)-Quinic acid, D-(-)-Quinic acid, D-Quinic acid, Quinic acid, Quinic acid','(-)-, [1R-(1α,3α,4α,5β)]-1,3,4,5-Tetrahydroxycyclohexanecarboxylic acid
InChI Formula InChI=1S/C7H12O6/c8-3-1-7(13,6(11)12)2-4(9)5(3)10/h3-5,8-10,13H,1-2H2,(H,11,12)/t3-,4-,5-,7+/m1/s1
SMILES O[C@@H]1C[C@@](O)(C[C@@H](O)[C@H]1O)C(=O)O
IUPAC Name (3R,5R)-1,3,4,5-tetrahydroxycyclohexane-1-carboxylic acid
Molecular Weight (g/mol) 192.17
Formula Weight 192.0634
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.