missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals D(+)-Raffinose pentahydrate, 99+%

Product Code. 10246730 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10246730 25 g Glass bottle 25g
10143830 100 g Glass bottle 100g
10618153 500 g Glass bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10246730 Supplier Thermo Scientific Chemicals Supplier No. 195670250
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 17629-30-0 | C18H32O16·5H2O | 594.52 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 17629-30-0
Molecular Formula C18H32O16·5H2O
Molecular Weight (g/mol) 594.52
MDL Number MFCD00071590
InChI Key BITMAWRCWSHCRW-ORBWWOJFSA-N
Synonym d-+-raffinosepentahydrate
PubChem CID 134129414
IUPAC Name (3S,4S,5R,6R)-2-[[(2R,3S,4S,5S)-6-[(2S,3S,4R,5R)-3,4-dihydroxy-2,5-bis(hydroxymethyl)oxolan-2-yl]oxy-3,4,5-trihydroxyoxan-2-yl]methoxy]-6-(hydroxymethyl)oxane-3,4,5-triol;pentahydrate
SMILES C(C1C(C(C(C(O1)OCC2C(C(C(C(O2)OC3(C(C(C(O3)CO)O)O)CO)O)O)O)O)O)O)O.O.O.O.O.O

Specifications

Melting Point 78.0°C to 80.5°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 99% min. (ELSD) (HPLC)
Packaging Glass bottle
Quantity 25 g
Beilstein 17, V,8, 403
Merck Index 14, 8096
Specific Rotation Condition + 105.00 (20.00°C c=10,water,dry basis)
Specific Rotation + 105.00
Solubility Information (10% in water) Clear colorless
Formula Weight 594.52
Percent Purity 99+%
Physical Form Crystalline Powder
Chemical Name or Material D(+)-Raffinose pentahydrate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.