missing translation for 'onlineSavingsMsg'
Learn More

D(+)-Raffinose pentahydrate, MedChemExpress

Product Code. 16409176
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16409176 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16409176 Supplier Medchem Express Supplier No. HYN1938
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

MedChemExpress D(+)-Raffinose pentahydrate (D-Raffinose pentahydrate) is a trisaccharide composed of galactose, glucose, and fructose that occurs naturally in a variety of vegetables and grains. D(+)-Raffinose pentahydrate is a functional oligosaccharide.

D(+)-Raffinose pentahydrate (D-Raffinose pentahydrate) is a trisaccharide composed of galactose, glucose, and fructose that occurs naturally in a variety of vegetables and grains. D(+)-Raffinose pentahydrate is a functional oligosaccharide.

Specifications

Chemical Name or Material D(+)-Raffinose pentahydrate
CAS 17629-30-0
Color White
Purity Grade Notes Research
Molecular Formula C18H42O21
Quantity 100 mg
Synonym D-Raffinose pentahydrate
Solubility Information DMSO : ≥ 100 mg/mL (168.21 mM)
SMILES OC[C@]1(O[C@H](CO)[C@@H](O)[C@@H]1O)O[C@H]([C@@H]([C@@H](O)[C@@H]2O)O)O[C@@H]2CO[C@H]([C@@H]([C@@H](O)[C@H]3O)O)O[C@@H]3CO.O.O.O.O.O
Molecular Weight (g/mol) 594.51
Formula Weight 594.51
Percent Purity 97.0%
Grade Research
Physical Form Powder
For Use With (Application) Metabolism-sugar/lipid metabolism
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.