missing translation for 'onlineSavingsMsg'
Learn More

D-(+)-Raffinose Pentahydrate, MP Biomedicals™

Product Code. 11465985 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
50 g
Unit Size:
50g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11465985 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11465985 Supplier MP Biomedicals Supplier No. 0210279750
 Restricted Item
Estimated Shipment: 22-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Composed of galactose, fructose and glucose. D-(+)-Raffinose Pentahydrate, MP Biomedicals is a trisaccharide and the most common alpha-galactosyl derivative of sucrose. Raffinose is hydrolysed to D-galactose and sucrose by D-galactosidase. This product is

Specifications

CAS 17629-30-0
Melting Point 76°C to 86°C
Density 1.465g/mL (Lit.)
Molecular Formula C18H42O21
Quantity 50 g
Synonym d-+-raffinosepentahydrate
Solubility Information Soluble in pyridine and water (1g/7mL).
InChI Key BITMAWRCWSHCRW-ORBWWOJFSA-N
Optical Rotation 101-107° (c=4, water)
SMILES C(C1C(C(C(C(O1)OCC2C(C(C(C(O2)OC3(C(C(C(O3)CO)O)O)CO)O)O)O)O)O)O)O.O.O.O.O.O
IUPAC Name (3S,4S,5R,6R)-2-[[(2R,3S,4S,5S)-6-[(2S,3S,4R,5R)-3,4-dihydroxy-2,5-bis(hydroxymethyl)oxolan-2-yl]oxy-3,4,5-trihydroxyoxan-2-yl]methoxy]-6-(hydroxymethyl)oxane-3,4,5-triol;pentahydrate
Molecular Weight (g/mol) 594.513
PubChem CID 134129414
Formula Weight 594.5
Percent Purity >98%
Physical Form Powder
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.