missing translation for 'onlineSavingsMsg'
Learn More

D-(+)-Raffinose Pentahydrate

Product Code. 15805188 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 g
500 g
1 kg
Packaging:
Plastic bottle
Unit Size:
100g
1kg
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15805188 100 g Plastic bottle 100g
15815188 500 g Plastic bottle 500g
15849762 1 kg Plastic bottle 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15805188 Supplier Thermo Scientific Supplier No. J21060.22
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 17629-30-0 | C18H42O21 | 594.513 g/mol

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 17629-30-0
Molecular Formula C18H42O21
Molecular Weight (g/mol) 594.513
MDL Number MFCD00071590
InChI Key BITMAWRCWSHCRW-ORBWWOJFSA-N
Synonym d-+-raffinosepentahydrate
PubChem CID 134129414
IUPAC Name (3 S,4 S,5 R,6 R)-2-[[(2 R,3 S,4 S,5 S)-6-[(2 S,3 S,4 R,5 R)-3,4-dihydroxy-2,5-bis(hydroxymethyl)oxolan-2-yl]oxy-3,4,5-trihydroxyoxan-2-yl]methoxy]-6-(hydroxymethyl)oxane-3,4,5-triol;pentahydrate
SMILES C(C1C(C(C(C(O1)OCC2C(C(C(C(O2)OC3(C(C(C(O3)CO)O)O)CO)O)O)O)O)O)O)O.O.O.O.O.O

Specifications

Boiling Point 78°C to 82°C
Packaging Plastic bottle
Quantity 100 g
Beilstein 4285763
Formula Weight 594.51
Physical Form Crystalline Powder
Chemical Name or Material D-(+)-Raffinose Pentahydrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.