missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals D-(-)-Ribose, 98%

Product Code. 11413508
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11413508 10 g 10g
11423508 50 g 50g
11433508 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11413508 Supplier Thermo Scientific Chemicals Supplier No. A17894.09
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 50-69-1 | C5H10O5 | 150.13 g/mol

D-(-)-Ribose is mainly used as a very basic source of metabolism of life. It is also used as a precursor for virazole, adenosine, thymidine, cytidine, fluoro-thymidine, 2-methyl adenosine, pyrromonazole toxina and adenosine methionine. It is further used in the synthesis of vitamin B2 (riboflavin), tetra-O-acetyl-ribose and nucleoside. It finds application as food additives and as supplement in cell culture.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
D-(-)-Ribose is mainly used as a very basic source of metabolism of life. It is also used as a precursor for virazole, adenosine, thymidine, cytidine, fluoro-thymidine, 2-methyl adenosine, pyrromonazole toxina and adenosine methionine. It is further used in the synthesis of vitamin B2 (riboflavin), tetra-O-acetyl-ribose and nucleoside. It finds application as food additives and as supplement in cell culture.

Solubility
Soluble in water. Insoluble in ether.

Notes
Hygroscopic. Incompatible with strong oxidizing agents. Store in a cool place.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 50-69-1
Molecular Formula C5H10O5
Molecular Weight (g/mol) 150.13
MDL Number MFCD00135453
InChI Key HMFHBZSHGGEWLO-ZHZWXSSZNA-N
Synonym 2r,3r,4r-2,3,4,5-tetrahydroxypentanal, aldehydo-d-ribose, ribose, d, d--ribose, aldehydo-d-ribo-pentose, ribo-2,3,4,5-tetrahydroxyvaleraldehyde, d, d-ribo-2,3,4,5-tetrahydroxyvaleraldehyde, ribose 9ci, dsstox_cid_23917, dsstox_gsid_43917
PubChem CID 5311110
ChEBI CHEBI:47014
IUPAC Name (2R,3R,4R)-2,3,4,5-tetrahydroxypentanal
SMILES OC[C@H]1OC(O)[C@H](O)[C@@H]1O

Specifications

Melting Point 82°C to 86°C
Assay Percent Range 0.98
Quantity 10 g
Beilstein 1723081
Sensitivity Hygroscopic
Merck Index 14,8204
Solubility Information Soluble in water. Insoluble in ether.
Optical Rotation −20° (c=10 in Water, 28h)
Formula Weight 150.13
Percent Purity 98%
Chemical Name or Material D-(-)-Ribose
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.