missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals D(-)-Ribose, 99+%

Product Code. 10605382 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
25g
500g
5g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10605382 100 g Glass bottle 100g
10320164 5 g Glass bottle 5g
10695372 25 g Glass bottle 25g
10629443 500 g Glass bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10605382 Supplier Thermo Scientific Chemicals Supplier No. 132361000
 Restricted Item
In Stock
Add to basket
Edge

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 50-69-1 | C5H10O5 | 150.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 50-69-1
Molecular Formula C5H10O5
Molecular Weight (g/mol) 150.13
MDL Number MFCD00135453
InChI Key HMFHBZSHGGEWLO-ZHZWXSSZNA-N
Synonym 2r,3r,4r-2,3,4,5-tetrahydroxypentanal, aldehydo-d-ribose, ribose, d, d--ribose, aldehydo-d-ribo-pentose, ribo-2,3,4,5-tetrahydroxyvaleraldehyde, d, d-ribo-2,3,4,5-tetrahydroxyvaleraldehyde, ribose 9ci, dsstox_cid_23917, dsstox_gsid_43917
PubChem CID 5311110
ChEBI CHEBI:47014
SMILES OC[C@H]1OC(O)[C@H](O)[C@@H]1O

Specifications

Melting Point 88.0°C to 92.0°C
Color Beige to White or Yellow
Loss on Drying 1% max. (40°C, 5 hrs, 20 torr)
Infrared Spectrum Authentic
Assay Percent Range 99% min. (ELSD) (HPLC)
Packaging Glass bottle
Quantity 100 g
Beilstein 01, I, 434
Merck Index 15, 8328
Specific Rotation Condition − 20.00 (20.00°C c=4,H2O)
Specific Rotation − 20.00
IUPAC Name (2R,3R,4R)-2,3,4,5-tetrahydroxypentanal
Formula Weight 150.13
Percent Purity 99+%
Physical Form Crystalline Powder
Chemical Name or Material D(-)-Ribose
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.