missing translation for 'onlineSavingsMsg'
Learn More

D-Ribose(mixture of isomers), MedChemExpress

Product Code. 16499396
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16499396 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16499396 Supplier Medchem Express Supplier No. HYW018772
 Restricted Item
Estimated Shipment: 24-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress D-Ribose(mixture of isomers) is an energy enhancer, and acts as a sugar moiety of ATP, and widely used as a metabolic therapy supplement for chronic fatigue syndrome or cardiac energy metabolism. D-Ribose(mixture of isomers) is active in protein glycation, induces NF-κB inflammation in a RAGE-dependent manner.

D-Ribose(mixture of isomers) is an energy enhancer, and acts as a sugar moiety of ATP, and widely used as a metabolic therapy supplement for chronic fatigue syndrome or cardiac energy metabolism. D-Ribose(mixture of isomers) is active in protein glycation, induces NF-κB inflammation in a RAGE-dependent manner.

Specifications

Chemical Name or Material D-Ribose(mixture of isomers)
CAS 50-69-1
Color Off-White
Purity Grade Notes Research
Molecular Formula C5H10O5
Quantity 100 mg
Solubility Information H2O : 100 mg/mL (666.09 mM; Need ultrasonic) ∣DMSO : ≥ 100 mg/mL (666.09 mM)
SMILES O=C[C@@H]([C@@H]([C@@H](CO)O)O)O
Molecular Weight (g/mol) 150.13
Formula Weight 150.13
Percent Purity 97.0%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-anti-virus
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.