missing translation for 'onlineSavingsMsg'
Learn More

D-(-)-Salicin, 99%

Product Code. 11487847
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11487847 100 g 100g
11467847 5 g 5g
11477847 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11487847 Supplier Thermo Scientific Alfa Aesar Supplier No. A16524.22
 Restricted Item
Estimated Shipment: 03-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 138-52-3 | C13H18O7 | 286.28 g/mol

Salicin is used as an analgesic and antipyretic. It is also used as a substrate for beta-glucosidase and an inhibitor of Cox. It finds application as an anti-inflammatory agent. It is involved in antipyretic action without causing gastric injury.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Soluble in water, alkalies, pyridine, glacial acetic acid and hot ethanol. Insoluble in ether and chloroform.

Notes
Store in cool place. Light and heat sensitive. Air senstive and moisture sensitive. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 138-52-3
Molecular Formula C13H18O7
Molecular Weight (g/mol) 286.28
MDL Number MFCD00006590
InChI Key NGFMICBWJRZIBI-UJPOAAIJSA-N
Synonym salicin, salicoside, salicine, salicyl alcohol glucoside, d---salicin, 2r,3s,4s,5r,6s-2-hydroxymethyl-6-2-hydroxymethyl phenoxy tetrahydro-2h-pyran-3,4,5-triol, saligenin beta-d-glucopyranoside, 2-hydroxymethyl phenyl beta-d-glucopyranoside, d-salicin, saligenin-beta-d-glucopyranoside
PubChem CID 439503
ChEBI CHEBI:17814
IUPAC Name (2R,3S,4S,5R,6S)-2-(hydroxymethyl)-6-[2-(hydroxymethyl)phenoxy]oxane-3,4,5-triol
SMILES C1=CC=C(C(=C1)CO)OC2C(C(C(C(O2)CO)O)O)O

Specifications

Melting Point 200°C to 203°C
Assay Percent Range 99%
Quantity 100 g
Beilstein 89593
Merck Index 14,8324
Solubility Information Soluble in water,alkalies,pyridine,glacial acetic acid and hot ethanol. Insoluble in ether and chloroform.
Formula Weight 286.28
Percent Purity 99%
Chemical Name or Material D-(-)-Salicin
compliance-icons
Product Identifier
  • D-(-)-Salicin
Signal Word
  • Warning
Hazard Category
  • Skin sensitiser Category 1
Hazard Statement
  • H317-May cause an allergic skin reaction.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
Supplemental information
  • MIXTURE LIST-Contain: D-(-)-Salicin

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.