missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals D-Serine, 99%

Product Code. 11430153
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11430153 25 g 25g
11420153 5 g 5g
11440153 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11430153 Supplier Thermo Scientific Chemicals Supplier No. A11353.14
 Restricted Item
Estimated Shipment: 08-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 312-84-5 | C3H7NO3 | 105.093 g/mol

D-Serine is involved in the biosynthesis of purines, pyrimidines, and other amino acids. D-Serine also is an agonist of glycine site of the NMDA-type glutamate receptor. It also acts as Lacosamide intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
D-Serine is involved in the biosynthesis of purines, pyrimidines, and other amino acids. D-Serine also is an agonist of glycine site of the NMDA-type glutamate receptor. It also acts as Lacosamide intermediate.

Solubility
Soluble in water (346 mg/ml at 20°C), and mineral acids. Insoluble in alcohol, ether, and benzene.

Notes
Store at 4°C. Store away from oxidizing agents. Keep container closed when not in use.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 312-84-5
Molecular Formula C3H7NO3
Molecular Weight (g/mol) 105.093
MDL Number MFCD00004269
InChI Key MTCFGRXMJLQNBG-UWTATZPHSA-N
Synonym d-serine, r-2-amino-3-hydroxypropanoic acid, 2r-2-amino-3-hydroxypropanoic acid, serine d-form, r-serine, serine, d, d-+-serine, h-d-ser-oh, d-2-amino-3-hydroxypropionic acid, d-serin
PubChem CID 71077
ChEBI CHEBI:16523
IUPAC Name (2R)-2-amino-3-hydroxypropanoic acid
SMILES C(C(C(=O)O)N)O

Specifications

Melting Point 218°C to 220°C (decomposition)
Assay Percent Range 99%
Beilstein 1721403
Merck Index 14,8460
Solubility Information Soluble in water (346mg/ml at 20°C),and mineral acids. Insoluble in alcohol,ether,and benzene.
Quantity 25 g
Formula Weight 105.09
Optical Rotation −13° (c=5 in 5N HCl)
Percent Purity 99%
Chemical Name or Material D-Serine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.