missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ D-Serine

Product Code. 30001064
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30001064 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”27950” to get your promotional price
Product Code. 30001064 Supplier Tocris Bioscience™ Supplier No. 0226/1G
  • ONLINE EXCLUSIVE PRICE
    150.00 €
    141.75 € / 1g
    Save 8.25 €
    5% Off
View all promotions
 Restricted Item
Estimated Shipment: 02-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”27950” to get your promotional price

Potentiator of NMDA recepors; acts at glycine site

D-Serine is a glycine agonist at the NMDA receptor.
TRUSTED_SUSTAINABILITY

Specifications

CAS 312-84-5
Quantity 1 g
Molecular Formula C3H7NO3
Formula Weight 105.09
Purity >98%
Molecular Weight (g/mol) 105.093
Chemical Name or Material D-Serine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.