missing translation for 'onlineSavingsMsg'
Learn More

D-Sorbitol, Molecular Biology Reagent, ≥98%, MP Biomedicals™

Product Code. 11496621 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
100 g
1 kg
5 kg
Unit Size:
100g
1kg
5kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11496621 5 kg 5kg
11476621 100 g 100g
11486621 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11496621 Supplier MP Biomedicals Supplier No. 0219485194
 Restricted Item
Estimated Shipment: 22-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

D-Sorbitol, Molecular Biology Reagent, ≥98% may be used to induce osmotic stress, and for washing spheroplasts and in isoelectric focusing to minimize endoosmotic flow in agarose gels.

  • D-Sorbitol is a sugar alcohol that is commonly used as a sugar substitute.
  • It occurs naturally and is also produced synthetically from glucose.
  • The food industry uses D-sorbitol as an additive in the form of a sweetener, humectant, emulsifier, thickener, or dietary supplement.
  • D-Sorbitol has also been found in cosmetics, paper, and pharmaceuticals.
  • Naturally, D-sorbitol occurs widely in plants via photosynthesis, ranging from algae to higher order fruits of the family Rosaceae.

Chemical Identifiers

CAS 50-70-4
Molecular Formula C6H14O6
Synonym d-sorbitol, sorbitol, d-glucitol, glucitol, l-gulitol, --sorbitol, diakarmon, glucarine, sorbol, d---sorbitol
PubChem CID 5780
ChEBI CHEBI:17924

Specifications

Melting Point 230°F
Color White
Density 1.49 g/cm3 at 59°F
pH 7.0
Boiling Point Very High
Flash Point 542°F
Quantity 5 kg
Solubility Information Water Solubility: 2,750,000 mg/L at 30°C
InChI Key FBPFZTCFMRRESA-JGWLITMVSA-N
Optical Rotation −2° ± 2° (c = 2, water)
SMILES C(C(C(C(C(CO)O)O)O)O)O
IUPAC Name (2R,3R,4R,5S)-hexane-1,2,3,4,5,6-hexol
Molecular Weight (g/mol) 182.172
Formula Weight 182.2
Vapor Pressure 9.9X10-9 mm Hg at 25°C
Percent Purity ≥98%
Grade Molecular Biology Grade
Physical Form Powder
Chemical Name or Material D-sorbitol
Show More Show Less

Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.