missing translation for 'onlineSavingsMsg'
Learn More
Learn More
D-(+)-Sucrose, crystals, BAKER ANALYZED™ ACS, J.T. Baker™
Shop All J.T.Baker Products
Click to view available options
:
1 kg
Description
- Meets ACS Reagent Chemical Requirements
- For Laboratory, Research, or Manufacturing Use
Specifications
Specifications
| CAS | 57-50-1 |
| Melting Point | 169°C to 170°C |
| Density | 1.29 g/cm³ (20°C) |
| Ignition Residue | ≤ 0.01% |
| Loss on Drying | ≤ 0.03% |
| Packaging | Plastic bottle for solids |
| Molecular Formula | C12H22O11 |
| MDL Number | MFCD00006626 |
| Quantity | 1 kg |
| Specific Rotation | 66.3 - 66.8 Degree |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction