missing translation for 'onlineSavingsMsg'
Learn More

D-(-)-Tartaric Acid, TRC

Product Code. 30423246
Change view
Click to view available options
Quantity:
1 g
25 g
100 g
Unit Size:
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30423246 25 g 25g
30417322 100 g -
30414953 1 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30423246 Supplier Toronto Research Chemicals Supplier No. TRCT00762525G
 Restricted Item
Estimated Shipment: 10-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 147-71-7
Molecular Formula C4 H6 O6
Molecular Weight (g/mol) 150.09
Synonym Butanedioic acid, 2,3-dihydroxy-, (2S,3S)-, Butanedioic acid, 2,3-dihydroxy-, [S-(R*,R*)]-, Tartaric acid, D-(-)- (8CI), (2S,3S)-2,3-Dihydroxybutanedioic acid, (-)-Tartaric acid, (2S,3S)-(-)-Tartaric acid, (2S,3S)-2,3-Dihydroxysuccinic acid, (2S,3S)-Tartaric acid, (S,S)-(-)-Tartaric acid, (S,S)-Tartaric acid, D-(-)-Tartaric acid, D-Tartaric acid, l-Tartaric acid, levo-Tartaric acid
IUPAC Name (2S,3S)-2,3-dihydroxybutanedioic acid
SMILES O[C@@H]([C@H](O)C(=O)O)C(=O)O

Specifications

Quantity 25 g
InChI Formula InChI=1S/C4H6O6/c5-1(3(7)8)2(6)4(9)10/h1-2,5-6H,(H,7,8)(H,9,10)/t1-,2-/m0/s1
Formula Weight 150.0164
Percent Purity >95%
Chemical Name or Material D-(-)-Tartaric Acid
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.