missing translation for 'onlineSavingsMsg'
Learn More

D(-)-Tartaric acid, 99%

Product Code. 10745801 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
250 g
1 kg
Packaging:
Glass Bottle
Plastic bottle
Unit Size:
100g
1kg
250g
25g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10745801 250 g Plastic bottle 250g
10306930 25 g Glass Bottle 25g
10224710 100 g Plastic bottle 100g
10794094 1 kg Plastic bottle 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10745801 Supplier Thermo Scientific Acros Supplier No. 137862500
 Restricted Item
Estimated Shipment: 09-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 147-71-7 | C4H6O6 | 150.09 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 147-71-7
Molecular Formula C4H6O6
Molecular Weight (g/mol) 150.09
MDL Number MFCD00004238
InChI Key FEWJPZIEWOKRBE-UHFFFAOYNA-N
Synonym d-tartaric acid, d---tartaric acid, 2s,3s-2,3-dihydroxysuccinic acid, d--tartaric acid, d-threaric acid, --d-tartaric acid, 2s,3s-2,3-dihydroxybutanedioic acid, --tartaric acid, s,s-tartaric acid, 2s,3s---tartaric acid
PubChem CID 439655
ChEBI CHEBI:15672
SMILES OC(C(O)C(O)=O)C(O)=O

Specifications

Melting Point 167°C to 171°C
Color White
Assay Percent Range 99%
Linear Formula HO2CCH(OH)CH(OH)CO2H
Beilstein 03, 520
Fieser 16,304
Merck Index 15, 9203
IUPAC Name (2S,3S)-2,3-dihydroxybutanedioic acid
Formula Weight 150.09
Percent Purity ≥98.5%
Quantity 250 g
Infrared Spectrum Authentic
Loss on Drying 0.5% max. (1 g, 105°C, 3 hrs)
Packaging Plastic bottle
Physical Form Crystals or Crystalline Powder
Chemical Name or Material D(-)-Tartaric acid, 99%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.