missing translation for 'onlineSavingsMsg'
Learn More

D-tert-Leucine, 99%

Product Code. 11398965
Change view
Click to view available options
Quantity:
100 mg
500 mg
2 g
Unit Size:
100mg
2g
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11398965 100 mg 100mg
11378965 500 mg 500mg
11388965 2 g 2g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11398965 Supplier Thermo Scientific Alfa Aesar Supplier No. L14098.MC
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 26782-71-8 | C6H13NO2 | 131.18 g/mol

D-tert-Leucine is used for synthesis of amino acids e.g. in the synthesis of aldehydes by double alkylation of unsaturated aldimines of tert-Leu esters.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
D-tert-Leucine is used for synthesis of amino acids e.g. in the synthesis of aldehydes by double alkylation of unsaturated aldimines of tert-Leu esters.

Solubility
Soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 26782-71-8
Molecular Formula C6H13NO2
Molecular Weight (g/mol) 131.18
MDL Number MFCD00004265
InChI Key NPDBDJFLKKQMCM-UHFFFAOYNA-N
Synonym d-tert-leucine, h-d-tle-oh, d-tert-butylglycine, r-2-amino-3,3-dimethylbutanoic acid, d-t-leucine, r-tert-leucine, 2r-2-amino-3,3-dimethylbutanoic acid, d-t-butylglycine, h-tbu-d-gly-oh, d-valine, 3-methyl
PubChem CID 6950340
IUPAC Name (2R)-2-amino-3,3-dimethylbutanoic acid
SMILES CC(C)(C)C(N)C(O)=O

Especificaciones

Melting Point >300°C
Beilstein 1721825
Solubility Information Soluble in water.
Quantity 100 mg
Formula Weight 131.18
Optical Rotation −7° (c=5 in 5N HCl)
Percent Purity 99%
Chemical Name or Material D-tert-Leucine

RUO – Research Use Only

Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.