missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals D-Threonine, 98%

Product Code. 10173531 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass Bottle
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10173531 25 g Glass Bottle 25g
10071781 5 g Glass Bottle 5g
10076233 100 g Glass Bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10173531 Supplier Thermo Scientific Chemicals Supplier No. 346970250
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

CAS: 632-20-2 | C4H9NO3 | 119.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemische Identifikatoren

CAS 632-20-2
Molecular Formula C4H9NO3
Molecular Weight (g/mol) 119.12
MDL Number MFCD00064269
InChI Key AYFVYJQAPQTCCC-STHAYSLISA-N
Synonym d-threonine, h-d-thr-oh, 2r,3s-2-amino-3-hydroxybutanoic acid, threonine, d, r-threonine, d-threonin, d-2-amino-3-hydroxybutyric acid, 2r,3s-2-amino-3-hydroxybutyric acid, d-thr, dth
PubChem CID 69435
ChEBI CHEBI:16398
IUPAC Name (2R,3S)-2-amino-3-hydroxybutanoic acid
SMILES C[C@H](O)[C@@H](N)C(O)=O

Spezifikation

Ammonium 200ppm max.
Arsenic (As) 1ppm max.
Color White
Chlorine (Cl) 200ppm max.
Heavy Metals (as Pb) 10ppm max.
Infrared Spectrum Authentic
Iron (Fe) 10ppm max.
Assay Percent Range 98%
Loss on Drying 0.3% max.
Other Amino Acids (TLC) Passes Test
Packaging Glass Bottle
Linear Formula CH3CH(OH)CH(NH)COH
Specific Rotation +27° to +29° (20°C, 589nm) (c=6, water)
Sulfate 300ppm max.
Transmittance 98% min. (c=5, H2O, 430nm) 1cm cell
Sulfated Ash 0.1% max.
Specific Rotation Condition +28° (20°C c=6,water)
Quantity 25 g
Formula Weight 119.12
Percent Purity ≥97.5%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material D-Threonine
Mehr anzeigen Weniger anzeigen

RUO – Research Use Only

Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.