missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals D-Trehalose, 99%, anhydrous

Product Code. 10125900 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Plastic bottle
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10125900 25 g Plastic bottle 25g
10308653 100 g Plastic bottle 100g
10677943 500 g Plastic bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10125900 Supplier Thermo Scientific Chemicals Supplier No. 309870250
 Restricted Item
Estimated Shipment: 20-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 99-20-7 | C12H22O11 | 342.30 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 99-20-7
Molecular Formula C12H22O11
Molecular Weight (g/mol) 342.3
MDL Number MFCD00006628
InChI Key HDTRYLNUVZCQOY-LIZSDCNHSA-N
Synonym trehalose, d-trehalose, alpha,alpha-trehalose, mycose, ergot sugar, alpha-d-trehalose, d-+-trehalose, d-+-trehalose, anhydrous, alpha,alpha'-trehalose, trehaose
PubChem CID 7427
ChEBI CHEBI:16551
IUPAC Name (2R,3S,4S,5R,6R)-2-(hydroxymethyl)-6-[(2R,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxyoxane-3,4,5-triol
SMILES C(C1C(C(C(C(O1)OC2C(C(C(C(O2)CO)O)O)O)O)O)O)O

Specifications

Melting Point 203.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (ELSD) on dry basis (HPLC)
Packaging Plastic bottle
Quantity 25 g
Merck Index 15,9742
Specific Rotation Condition +197.50 (20.00°C c=2,H2O)
Specific Rotation + 197.50
Solubility Information (10% in water) Clear colorless to light yellow
Formula Weight 342.3
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material D-Trehalose, Anhydrous
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.