missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Melford D-(+)-Trehalose Dihydrate
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
Description
Product usually ships within 24 hours. Research or further manufacturing use only, not for food or drug use.
- CAS Number: 6138-23-4
- Molecular Formula: C12H22O11 2H2O
- Molecular Weight: 378.3
Specifications
Specifications
| CAS | 6138-23-4 |
| Molecular Formula | C12H22O11-2H2O |
| Quantity | 25 g |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction