missing translation for 'onlineSavingsMsg'
Learn More

D-(+)-Trehalose, Dihydrate, Fisher BioReagents

Product Code. 10744344
Change view
Click to view available options
Quantity:
10 g
25 g
100 g
1 kg
Packaging:
Poly Bottle
Unit Size:
100g
10g
1kg
25g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10744344 100 g Poly Bottle 100g
11403104 10 g Poly Bottle 10g
11413104 25 g Poly Bottle 25g
10318094 1 kg Poly Bottle 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10744344 Supplier Fisher BioReagents Supplier No. BP2687100
Spotlight Offers
Use promo code 28038
View all promotions
 Restricted Item
Estimated Shipment: 18-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
D-(+)-Trehalose is used in several biochemical techniques.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6138-23-4
Molecular Formula C12H26O13
Molecular Weight (g/mol) 378.327
MDL Number MFCD00071594
InChI Key DPVHGFAJLZWDOC-PVXXTIHASA-N
Synonym trehalose dihydrate, d-+-trehalose dihydrate, a,a-trehalose, d +-trehalose dihydrate, unii-7yin7j07x4, alpha,alpha-trehalose dihydrate, d-trehalose dihydrate, mycose,1-o-alpha-d-glucopyranosyl-alpha-d-glucopyranoside, alp,alp.-trehalose, 2r,3s,4s,5r,6r-2-hydroxymethyl-6-2r,3r,4s,5s,6r-3,4,5-trihydroxy-6-hydroxymethyl tetrahydropyran-2-yl oxy-tetrahydropyran-3,4,5-triol
PubChem CID 181978
IUPAC Name (2R,3S,4S,5R,6R)-2-(hydroxymethyl)-6-[(2R,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxyoxane-3,4,5-triol;dihydrate
SMILES C(C1C(C(C(C(O1)OC2C(C(C(C(O2)CO)O)O)O)O)O)O)O.O.O

Specifications

Melting Point 97°C
Color White
Loss on Drying 8.5 to 10.5%
Infrared Spectrum Conforms to reference
Assay Percent Range ≥98 %
Packaging Poly Bottle
Quantity 100 g
Merck Index 15, 9742
Formula Weight 378.33
Percent Purity 98% min.
Physical Form Powder or Solid
Chemical Name or Material D-(+)-Trehalose, Dihydrate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.