missing translation for 'onlineSavingsMsg'
Learn More

D-(+)-Trehalose Dihydrate, MP Biomedicals

Product Code. 11453491 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
100 g
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11453491 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11453491 Supplier MP Biomedicals Supplier No. 0210309780
  • ONLINE EXCLUSIVE PRICE
    486.00 €
    460.95 € / 100g
    Save 25.05 €
    5% Off
 Restricted Item
Estimated Shipment: 24-02-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Natural alpha-linked disaccharide used as a protein-stabilizing agent and cryoprotectant in cell freezing media.

  • Trehalose does not reduce Fehling's solution; it is fermented by yeast
  • It is not split by alpha-glucosidase
  • Acid hydrolysis gives 2 mols D-glucose

Specifications

CAS 6138-23-4
Melting Point 203°C (literature)
Color White
Molecular Formula C12H26O13
Linear Formula C12H22O11 . 2H2O
MDL Number MFCD00071594
Quantity 100 g
Synonym trehalose dihydrate, d-+-trehalose dihydrate, a,a-trehalose, d +-trehalose dihydrate, unii-7yin7j07x4, alpha,alpha-trehalose dihydrate, d-trehalose dihydrate, mycose,1-o-alpha-d-glucopyranosyl-alpha-d-glucopyranoside, alp,alp.-trehalose, 2r,3s,4s,5r,6r-2-hydroxymethyl-6-2r,3r,4s,5s,6r-3,4,5-trihydroxy-6-hydroxymethyl tetrahydropyran-2-yl oxy-tetrahydropyran-3,4,5-triol
Solubility Information Soluble in water (50mg/mL - clear, colorless solution) or hot alcohol.
InChI Key DPVHGFAJLZWDOC-PVXXTIHASA-N
SMILES C(C1C(C(C(C(O1)OC2C(C(C(C(O2)CO)O)O)O)O)O)O)O.O.O
IUPAC Name (2R,3S,4S,5R,6R)-2-(hydroxymethyl)-6-[(2R,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxyoxane-3,4,5-triol;dihydrate
Molecular Weight (g/mol) 378.327
PubChem CID 181978
Formula Weight 378.3
Percent Purity ≥98%
Physical Form Powder
Chemical Name or Material D-(+)-Trehalose Dihydrate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.