missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals D(+)-Tryptophan, 99%

Product Code. 10627104 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
Unit Size:
100g
25g
500g
5g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10627104 100 g 100g
10367680 5 g 5g
10533341 25 g 25g
11966611 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10627104 Supplier Thermo Scientific Chemicals Supplier No. 168631000
 Restricted Item
Estimated Shipment: 12-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 153-94-6 | C11H12N2O2 | 204.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 153-94-6
Molecular Formula C11H12N2O2
Molecular Weight (g/mol) 204.23
MDL Number MFCD00005647
InChI Key QIVBCDIJIAJPQS-SECBINFHSA-N
Synonym d-tryptophan, h-d-trp-oh, r-tryptophan, d +-tryptophan, d-trytophane, d-tryptophane, tryptophan, d, 2r-2-amino-3-1h-indol-3-yl propanoic acid, +-tryptophan, h-d-typ-oh
PubChem CID 9060
ChEBI CHEBI:16296
IUPAC Name (2R)-2-amino-3-(1H-indol-3-yl)propanoic acid
SMILES N[C@H](CC1=CNC2=CC=CC=C12)C(O)=O

Specifications

Melting Point 282.0°C to 285.0°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 99%
Loss on Drying 0.5% max.
Packaging Glass Bottle
Specific Rotation + 31.50
Solubility Information Solubility in water: 11g/L (20°C). Other solubilities: soluble in alkali hydroxides, soluble in hot alcohol, insoluble in chloroform
Specific Rotation Condition + 31.50 (24.00°C c=1,H2O)
Quantity 100 g
Formula Weight 204.23
Percent Purity 99%
Physical Form Powder
Chemical Name or Material D(+)-Tryptophan
Show More Show Less

RUO – Research Use Only

Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.