missing translation for 'onlineSavingsMsg'
Learn More

D-Valine, 99%, MP Biomedicals™

Product Code. 11497441 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11497441 5 g 5g
11407451 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11497441 Supplier MP Biomedicals Supplier No. 0210322605
 Restricted Item
Estimated Shipment: 29-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 640-68-6
Molecular Formula C5H11NO2
Molecular Weight (g/mol) 117.15
MDL Number MFCD00064219
InChI Key KZSNJWFQEVHDMF-SCSAIBSYSA-N
Synonym d-valine, h-d-val-oh, r-valine, valine, d, d-val, 2r-2-amino-3-methylbutanoic acid, d-2-aminoisovaleric acid, d-valin, unii-y14i1443ur, d-2-amino-3-methylbutanoic acid
PubChem CID 71563
ChEBI CHEBI:27477
IUPAC Name (2R)-2-amino-3-methylbutanoic acid
SMILES CC(C)[C@@H](N)C(O)=O

Specifications

Assay Percent Range 99%
Quantity 5 g
Formula Weight 117.1
Percent Purity ≥98%
Physical Form Solid
Chemical Name or Material D-Valine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.