missing translation for 'onlineSavingsMsg'
Learn More

D-Valine methyl ester hydrochloride, 99%

Product Code. 10746115 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass Bottle
Unit Size:
1g
5g
Product Code. Quantity Packaging unitSize
10746115 1 g Glass Bottle 1g
10386874 5 g Glass Bottle 5g
2 options
This item is not returnable. View return policy

Product Code. 10746115

Brand: Thermo Scientific Acros 398720010

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 7146-15-8 | C6H14ClNO2 | 167.63 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7146-15-8
Molecular Formula C6H14ClNO2
Molecular Weight (g/mol) 167.63
MDL Number MFCD00237309
InChI Key KUGLDBMQKZTXPW-NUBCRITNSA-N
Synonym d-valine methyl ester hydrochloride, methyl d-valinate hydrochloride, h-d-val-ome.hcl, r-methyl 2-amino-3-methylbutanoate hydrochloride, d-valinemethyl ester hydrochloride, methyl 2r-2-amino-3-methylbutanoate hydrochloride, d-valine, methyl ester, hydrochloride, h-d-val-ome, methyl d-valinate hcl, h-d-val-ome hcl
PubChem CID 11984198
SMILES Cl.COC(=O)[C@H](N)C(C)C

Specifications

Color White
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass Bottle
Linear Formula (CH3)2CHCH(NH2)COOCH3·HCl
Specific Rotation -13.5° to -17.5° (20°C, 589nm) (c=2, H2O)
IUPAC Name methyl (2R)-2-amino-3-methylbutanoate;hydrochloride
Quantity 1 g
Formula Weight 167.63
Percent Purity ≥98.5%
Physical Form Powder
Chemical Name or Material D-Valine methyleester hydrochloride
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.