missing translation for 'onlineSavingsMsg'
Learn More

D-(-)-Valinol, 98%

Product Code. 11319025
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11319025 1 g 1g
11329025 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11319025 Supplier Thermo Scientific Alfa Aesar Supplier No. L14166.03
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 4276-09-9 | C5H13NO | 103.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4276-09-9
Molecular Formula C5H13NO
Molecular Weight (g/mol) 103.17
MDL Number MFCD00064297
InChI Key NWYYWIJOWOLJNR-YFKPBYRVSA-N
Synonym d-valinol, r---2-amino-3-methyl-1-butanol, r-2-amino-3-methylbutan-1-ol, 2r-2-amino-3-methylbutan-1-ol, r-2-amino-3-methyl-1-butanol, d-2-amino-3-methyl-1-butanol, 2r-2-amino-3-methyl-1-butanol, 1r-1-isopropyl-2-hydroxyethylamine, d---valinol r---2-amino-3-methyl-1-butanol, d-valinol r---2-amino-3-methyl-1-butanol
PubChem CID 6950587
IUPAC Name (2R)-2-amino-3-methylbutan-1-ol
SMILES CC(C)[C@@H](N)CO

Specifications

Refractive Index 1.455
Density 0.931
Boiling Point 189°C to 190°C
Melting Point 30°C to 34°C
Quantity 1 g
Flash Point 78°C (172°F)
Optical Rotation −11° (c=10 in Water)
Sensitivity Air sensitive
Beilstein 1719138
Formula Weight 103.17
Percent Purity 98%
Chemical Name or Material D-(-)-Valinol
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.