missing translation for 'onlineSavingsMsg'
Learn More

D-(+)-Xylose, MP Biomedicals™

Product Code. 11427531 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
1 kg
Unit Size:
1kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11427531 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11427531 Supplier MP Biomedicals Supplier No. 0210330091
 Restricted Item
Estimated Shipment: 17-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Classified as a monosaccharide of the aldopentose type. D-(+)-Xylose, MP Biomedicals is a sugar first isolated from wood. It contains five carbon atoms and includes a formyl functional group. Xylose is the main building block for hemicellulose.

  • Xylose is the main building block for hemicellulose
  • The acid-catalysed degradation of hemicellulose gives furfural, a specialty solvent in industry and a precursor to synthetic polymers
  • In animal medicine, used to test for malabsorption by administration in water after fasting

Specifications

CAS 58-86-6
Melting Point 155°C to 157°C (literature)
Molecular Formula C5H10O5
Quantity 1 kg
Synonym Wood sugar
Solubility Information Soluble in water (clear,colorless solution), pyridin, and hot alcohol.
Formula Weight 150.13
Physical Form Powder
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.