missing translation for 'onlineSavingsMsg'
Learn More

D-(+)-Xylose (White to Off-White Powder), Fisher BioReagents

Product Code. 10572945
Change view
Click to view available options
Quantity:
250 g
Packaging:
Glass Bottle
Unit Size:
250g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10572945 250 g Glass Bottle 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10572945 Supplier Fisher BioReagents Supplier No. BP708250
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
D-(+)-Xylose is often used as a cell culture additive.
TRUSTED_SUSTAINABILITY

Specifications

Alpha Vector Xylose
CAS 58-86-6
Melting Point 148°C
Color White
Assay Percent Range ≥98 %
Packaging Glass Bottle
Molecular Formula C5H10O5
Quantity 250 g
Merck Index 15, 10285
Specific Rotation 19 to 21° (+ or -)
Formula Weight 150.13
Percent Purity ≥98%
Physical Form Powder or Solid
Chemical Name or Material D-(+)-Xylose
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.