missing translation for 'onlineSavingsMsg'
Learn More

D-Xylulose (0.5 M in Water), TRC

Product Code. 30452215
Change view
Click to view available options
Quantity:
5 mg
25 mg
100 mg
Unit Size:
100mg
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30452215 25 mg 25mg
30456576 100 mg 100mg
30453707 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30452215 Supplier Toronto Research Chemicals Supplier No. TRCX75090025MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 551-84-8
Molecular Formula C5 H10 O5
Molecular Weight (g/mol) 150.13
Synonym D-threo-2-Pentulose, D-threo-Pentulose (7CI,8CI), D-Xylulose, Xylulose
IUPAC Name (3S,4R)-1,3,4,5-tetrahydroxypentan-2-one
SMILES OC[C@@H](O)[C@H](O)C(=O)CO

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C5H10O5/c6-1-3(8)5(10)4(9)2-7/h3,5-8,10H,1-2H2/t3-,5+/m1/s1
Formula Weight 150.0528
Concentration or Composition (by Analyte or Components) 0.5 M
Chemical Name or Material D-Xylulose
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.