missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ DAB Quanto Detection System

Product Code. 12623957 Shop All Epredia Products
Click to view available options
Quantity:
1 L
125 mL DAB Quanto Substrate, 4 mL DAB Quanto Chromagen
60 mL DAB Quanto Substrate, 2 mL DAB Quanto Chromagen
Unit Size:
1 L
125mL
60mL
This item is not returnable. View return policy
DAB Quanto Chromogen contains Diaminobenzidine (DAB) in buffer. DAB is a suspected carcinogen. Do not pipet by mouth. Avoid contact with skin and eyes. Reagent is acidic and can cause burns if skin contact occurs. Handle with care and dispose of according to regulations. DAB Quanto Substrate contains Hydrogen Peroxide in buffer. Avoid contact with skin and eyes.

Product Code. 12623957

Brand: Epredia™ TA060QHDX

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
DAB Quanto Chromogen contains Diaminobenzidine (DAB) in buffer. DAB is a suspected carcinogen. Do not pipet by mouth. Avoid contact with skin and eyes. Reagent is acidic and can cause burns if skin contact occurs. Handle with care and dispose of according to regulations. DAB Quanto Substrate contains Hydrogen Peroxide in buffer. Avoid contact with skin and eyes.

Provides a strong brown staining and can be used after reconstitution for up to 1 week at room temperature and up to 2 weeks at 2° to 8°C.

DAB Quanto has been validated in combination with both UltraVision LP and UltraVision Quanto and provides four major benefits.
    • Strong staining—ideal to quantifying cells and protein expression
    • Less mixing—reducing personnel time and stress
    • Less disposal—cuts down waste
    • Lower risk of mixing errors—supports higher staining consistency
    3,3' Diaminobenzidine (DAB) is a widely used chromogen for immunohistochemical staining and immunoblotting. The DAB Quanto is an entirely new formula for a diaminobenzidine-based peroxidase substrate that develops faster on tissues sections reducing incubation times. It utilizes novel chemistry to provide greater sensitivity, generating a crisper, darker, insoluble precipitate. The standard working dilution is 30μL of DAB Quanto Chromogen per 1ml of DAB Quanto Substrate. The use of liquid components reduces some risks associated with the handling of powders or tablets. The DAB Quanto offers an added advantage, as the working solution is stable for up to 2 weeks at 2 to 8°C and up to a week at room temperature enabling continued use and minimizing waste disposal.
    • Supplied in a two-component form consisting of a DAB Quanto Chromogen and DAB Quanto Substrate
    • Standard working dilution is 30μL of DAB Quanto Chromogen per 1mL of DAB Quanto Substrate
    • Use of liquid components reduces some risks associated with the handling of powders or tablets
  • TRUSTED_SUSTAINABILITY

    Specifications

    Chromagen Type DAB Quanto Chromagen
    Content And Storage 2° to 8°C
    Quantity 60 mL DAB Quanto Substrate, 2 mL DAB Quanto Chromagen
    Substrate Type Dab Quanto Subrate
    compliance-icons
    Product Identifier
    • DAB Quanto (HRP) Detection System Substrate
    Signal Word
    • Danger
    Hazard Category
    • Carcinogenicity Category 2
    • Germ cell mutagenicity Category 2
    • Skin corrosion/irritation Category 1 B
    Hazard Statement
    • H341-Suspected of causing genetic defects.
    • H351-Suspected of causing cancer.
    • H314-Causes severe skin burns and eye damage.
    Precautionary Statement
    • P201-Obtain special instructions before use.
    • P202-Do not handle until all safety precautions have been read and understood.
    • P405-Store locked up.
    • P501-Dispose of contents/container to location in accordance with local/regional/national/international regulations.
    • P308+P313-IF exposed or concerned: Get medical advice/attention.
    Product Content Correction

    Your input is important to us. Please complete this form to provide feedback related to the content on this product.

    Product Title

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

    Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.