missing translation for 'onlineSavingsMsg'
Learn More

Dabigatran, MedChemExpress

Product Code. 16484656
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16484656

Brand: Medchem Express HY10163

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Dabigatran (BIBR 953), an oral anticoagulant, is a reversible, potent, competitive direct thrombin inhibitor (Ki=4.5 nM). Dabigatran (BIBR 953) also inhibits thrombin-induced platelet aggregation (IC50=10 nM).

Dabigatran (BIBR 953), an oral anticoagulant, is a reversible, potent, competitive direct thrombin inhibitor (Ki=4.5 nM). Dabigatran (BIBR 953) also inhibits thrombin-induced platelet aggregation (IC50=10 nM).

Specifications

Chemical Name or Material Dabigatran
CAS 211914-51-1
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C25H25N7O3
Quantity 100 mg
Synonym BIBR 953 BIBR 953ZW
Solubility Information 0.1 M HCL : 12.5 mg/mL (26.51 mM; Need ultrasonic) ∣H2O : < 0.1 mg/mL (insoluble) ∣DMSO : < 1 mg/mL (insoluble or slightly soluble)
SMILES N=C(N)C(C=C1)=CC=C1NCC2=NC3=CC(C(N(CCC(O)=O)C4=NC=CC=C4)=O)=CC=C3N2C
Molecular Weight (g/mol) 471.51
Formula Weight 471.51
Percent Purity 96.84%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.