missing translation for 'onlineSavingsMsg'
Learn More

Dabsyl Chloride, TRC

Product Code. 30354844
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30354844 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30354844 Supplier Toronto Research Chemicals Supplier No. TRCD101100250MG
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifica

Chemical Name or Material Dabsyl Chloride
CAS 56512-49-3
Molecular Formula C14 H14 Cl N3 O2 S
Quantity 250 mg
Synonym 4-[[4-(Dimethylamino)phenyl]azo]benzenesulfonyl Chloride, 4-Dimethylaminoazobenzene-4'-sulfonyl Chloride, 4'-(Dimethylamino)azobenzene-4-sulfonic Acid Chloride, DABS-Cl
InChI Formula InChI=1S/C14H14ClN3O2S/c1-18(2)13-7-3-11(4-8-13)16-17-12-5-9-14(10-6-12)21(15,19)20/h3-10H,1-2H3/b17-16+
SMILES CN(C)c1ccc(cc1)N=Nc2ccc(cc2)S(=O)(=O)Cl
IUPAC Name 4-[(E)-[4-(dimethylamino)phenyl]azo]benzenesulfonyl chloride
Molecular Weight (g/mol) 323.8
Formula Weight 323.0495
Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.