missing translation for 'onlineSavingsMsg'
Learn More

Dabsyl Chloride, TRC

Product Code. 30365243
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30365243 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30365243 Supplier Toronto Research Chemicals Supplier No. TRCD1011001G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Dabsyl Chloride
CAS 56512-49-3
Molecular Formula C14 H14 Cl N3 O2 S
Quantity 1 g
Synonym 4-[[4-(Dimethylamino)phenyl]azo]benzenesulfonyl Chloride, 4-Dimethylaminoazobenzene-4'-sulfonyl Chloride, 4'-(Dimethylamino)azobenzene-4-sulfonic Acid Chloride, DABS-Cl
InChI Formula InChI=1S/C14H14ClN3O2S/c1-18(2)13-7-3-11(4-8-13)16-17-12-5-9-14(10-6-12)21(15,19)20/h3-10H,1-2H3/b17-16+
SMILES CN(C)c1ccc(cc1)N=Nc2ccc(cc2)S(=O)(=O)Cl
IUPAC Name 4-[(E)-[4-(dimethylamino)phenyl]azo]benzenesulfonyl chloride
Molecular Weight (g/mol) 323.8
Formula Weight 323.0495
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.