missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Daclatasvir dihydrochloride, MedChemExpress
Description
Daclatasvir dihydrochloride (BMS-790052 dihydrochloride) is a potent and orally active HCV NS5A protein inhibitor with EC50s range of 9-146 pM for multiple HCV replicon genotypes. Daclatasvir dihydrochloride is also an organic anion transporting polypeptide 1B (OATP1B) and OATP1B3 inhibitor with IC50s of 1.5 μM and 3.27 μM, respectively.
Specifications
Specifications
| Chemical Name or Material | Daclatasvir dihydrochloride |
| CAS | 1009119-65-6 |
| Color | White |
| Purity Grade Notes | Research |
| Molecular Formula | C40H52Cl2N8O6 |
| Quantity | 5 mg |
| Synonym | BMS-790052 dihydrochlorideEBP 883 dihydrochloride |
| Solubility Information | DMSO : ≥ 56 mg/mL (68.98 mM) ∣H2O : 50 mg/mL (61.59 mM; Need ultrasonic) |
| SMILES | O=C(N1CCC[C@H]1C2=NC=C(C3=CC=C(C4=CC=C(C=C4)C5=CN=C(N5)[C@@H]6CCCN6C([C@H](C(C)C)NC(OC)=O)=O)C=C3)N2)[C@H](C(C)C)NC(OC)=O.[H]Cl.[H]Cl |
| Molecular Weight (g/mol) | 811.8 |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?