missing translation for 'onlineSavingsMsg'
Learn More

Dacthal, TRC

Product Code. 30353262
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30353262 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30353262 Supplier Toronto Research Chemicals Supplier No. TRCD1046401G
 Restricted Item
Estimated Shipment: 14-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

CAS 1861-32-1
Molecular Formula C10 H6 Cl4 O4
Quantity 1 g
Synonym 1,4-Benzenedicarboxylic acid, 2,3,5,6-tetrachloro-, dimethyl ester (9CI), Terephthalic acid, tetrachloro-, dimethyl ester (6CI,8CI), 2,3,5,6-Tetrachloro-1,4-benzenedicarboxylic acid dimethyl ester, Cepos, Chlorthal dimethyl ester, Chlorthal-dimethyl, Chlorthal-methyl, DAC 893, DCPA, DCPA (herbicide), Dac 4, Dacthal, Dimethyl 2,3,5,6-tetrachloroterephthalate, Dimethyl tetrachloroterephthalate, NSC 155745, TCTP, Tetrachloroterephthalic acid dimethyl ester, Tetral
InChI Formula InChI=1S/C10H6Cl4O4/c1-17-9(15)3-5(11)7(13)4(10(16)18-2)8(14)6(3)12/h1-2H3
SMILES COC(=O)c1c(Cl)c(Cl)c(C(=O)OC)c(Cl)c1Cl
IUPAC Name dimethyl 2,3,5,6-tetrachlorobenzene-1,4-dicarboxylate
Molecular Weight (g/mol) 331.96
Formula Weight 329.902
Chemical Name or Material Chlorthal-dimethyl
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.