missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sharpsguard™ Dani 22 Waste Container

Click to view available options
Capacity (Metric):
21.01 L
Unit Size:
Each
Description
- Sharpsguard™ eco dani 22 is for the safe collection and transportation of broken glass and crockery.
- Color-coded with a solid orange lid, handle and label, making it easy to distinguish from other waste containers.
- It should be disposed of in landfill sites and therefore is not suitable for hazardous or infectious items.
Specifications
Specifications
| Capacity (Metric) | 21.01 L |
| Diameter (Metric) Outer | 324 mm |
| Color | Orange |
| Height (Metric) | 343 mm |
| Quantity | 1 Ea. |
| For Use With (Application) | Disposal of Broken Glass and Crockery |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction