missing translation for 'onlineSavingsMsg'
Learn More

Trajan Scientific and Medical™ DANI Liners

Product Code. 15269043
Change view
Click to view available options
Description:
Single Taper
Straight-through Liner
Straight-through Liner w/ Quartz Wool
Tapered Focus Liner
Unit Size:
Pack of 5
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Description Unit Size
15269043 Tapered Focus Liner Pack of 5
15249043 Single Taper Pack of 5
15219043 Straight-through Liner Pack of 5
15209043 Straight-through Liner w/ Quartz Wool Pack of 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 15269043 Supplier Trajan Scientific and Medical™ Supplier No. 092506
Estimated Shipment: 06-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Provides better analysis for a wide range of compounds. Trajan Scientific and Medical DANI Liners has unique deactivation process for SGE standard inlets.

  • Each batch of liners tested for activity
  • Manufactured from borosilicate glass
  • Color coded by geometry for easy identification
  • For use with DANI instruments

Specifications

Product Type DANI Liners
Description Tapered Focus Liner
For Use With (Application) Deactivated Liners For DANI GCs
Quantity 5/Pk.
Length (Metric) 70 mm
Diameter (Metric) Inner 4 mm
Diameter (Metric) Outer 6 mm
For Use With (Equipment) Tapered FocusLiner for DANI
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.