missing translation for 'onlineSavingsMsg'
Learn More

MedChemExpress Dansyl chloride

Product Code. 16498876
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16498876 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16498876 Supplier MedChemExpress Supplier No. HYD0017

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

MedChemExpress Dansyl chloride is a reagent that reacts with primary amino groups in both aliphatic and aromatic amines to produce stable blue- or blue-green-fluorescent sulfonamide adducts.

Specifications

Content And Storage 4°C, stored under nitrogen ∣ The compound is unstable in solutions, freshly prepared is recommended.
Product Type Dye Reagents
Quantity 100 mg
Shipping Condition Room temperature
CAS 605-65-2
Molecular Weight (g/mol) 269.75 g/mol
Synonym DNSCl
Target HBV
Solubility DMSO : 6.67 mg/mL (24.73 mM; Need ultrasonic)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.