missing translation for 'onlineSavingsMsg'
Learn More

Dansyl chloride, 98%

Product Code. 10540571 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10540571 1 g Glass bottle 1g
10070880 5 g Glass bottle 5g
10348743 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10540571 Supplier Thermo Scientific Acros Supplier No. 115850010
 Restricted Item
Estimated Shipment: 09-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 605-65-2 | C12H12ClNO2S | 269.74 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 605-65-2
Molecular Formula C12H12ClNO2S
Molecular Weight (g/mol) 269.74
MDL Number MFCD00003985
InChI Key XPDXVDYUQZHFPV-UHFFFAOYSA-N
Synonym 5-Dimethylamino-1-naphthalenesulfonyl chloride
PubChem CID 11801
ChEBI CHEBI:51907
IUPAC Name 5-(dimethylamino)naphthalene-1-sulfonyl chloride
SMILES CN(C)C1=CC=CC2=C(C=CC=C12)S(Cl)(=O)=O

Specifications

Melting Point 71.0°C to 74.0°C
Infrared Spectrum Authentic
Packaging Glass bottle
Assay Percent Range 98%
Color Beige/Brown or Gray to White
Solubility Information Solubility in water: 10g/L (20°C). Other solubilities: soluble in acetone, pyridine, benzene and dioxane
Quantity 1 g
Physical Form Powder, Needles or Crystals
Formula Weight 269.75
Percent Purity 98%
Chemical Name or Material Dansyl chloride, 98%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Dansyl chloride
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 1 B
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H302-Harmful if swallowed.
  • H314-Causes severe skin burns and eye damage.
  • EUH029-Contact with water liberates toxic gas.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P402+P404-Store in a dry place. Store in a closed container.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.